missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-methylphenol, 98%, Thermo Scientific™

Product Code. 11359925
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-70-0
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00042296
InChI Key RVYGYYVGWSCWGY-UHFFFAOYSA-N
Synonym 4-fluoro-m-cresol, 3-methyl-4-fluorophenol, 2-fluoro-5-hydroxytoluene, 4-fluoro-3-methyl-phenol, phenol, 4-fluoro-3-methyl, 4-fluoro-3-methylphenol, pubchem1501, 3-methyl-4-fluoro phenol, 4-fluoro-3-methyl phenol, acmc-209k1i
PubChem CID 2737394
IUPAC Name 4-fluoro-3-methylphenol
SMILES CC1=C(C=CC(=C1)O)F

Specifications

Melting Point 28°C to 32°C
Density 1.134
Boiling Point 76°C (5 mmHg)
Flash Point 96°C (204°F)
Odor Phenol-like
Refractive Index 1.515
Quantity 25 g
UN Number UN3261
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-methylphenol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'