missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

37.10 € - 119.00 €

Chemical Identifiers

CAS 453-71-4
Molecular Formula C7H4FNO4
Molecular Weight (g/mol) 185.11
MDL Number MFCD00007058
InChI Key BOJWTAQWPVBIPG-UHFFFAOYSA-N
Synonym 4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
PubChem CID 67987
IUPAC Name 4-fluoro-3-nitrobenzoic acid
SMILES OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10430921
View Documents Promotion Details
Thermo Scientific Acros
305060050
5 g Glass bottle
37.10 €
5g
Please to purchase this item. Need a web account? Register with us today!
10783583
View Documents Promotion Details
Thermo Scientific Acros
305060250
25 g Glass bottle
119.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

453-71-4
185.11
BOJWTAQWPVBIPG-UHFFFAOYSA-N
67987
OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
C7H4FNO4
MFCD00007058
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
4-fluoro-3-nitrobenzoic acid

Specifications

453-71-4
100.0
Tan to Yellow
97.5% min. (GC)
FC6H3(NO2)CO2H
4-fluoro-3-nitrobenzoic acid, 3-nitro-4-fluorobenzoic acid, p-fluoro-3-nitrobenzoic acid, 4-fluoro-3-nitrobenzoicacid, 5-carboxy-2-fluoronitrobenzene, benzoic acid, 4-fluoro-3-nitro, 4-fluoro-3-nitro-benzoic acid, pubchem1306, acmc-1aqvm, 3-nitro-4-flurobenzoic acid
BOJWTAQWPVBIPG-UHFFFAOYSA-N
4-fluoro-3-nitrobenzoic acid
67987
98%
4-Fluoro-3-nitrobenzoic acid, 98%
97.5
120.0°C to 126.0°C
Authentic
C7H4FNO4
MFCD00007058
Solubility in water: insoluble in water
OC(=O)C1=CC=C(F)C(=C1)[N+]([O-])=O
185.11
185.11
Crystals or Powder
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 207-221-3

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only