missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-7-nitrobenzofurazan, Thermo Scientific Chemicals Product Code.: 15483939

4-Fluoro-7-nitrobenzofurazan, Thermo Scientific Chemicals

Product Code. 15483939
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 15483939

Brand: Thermo Scientific Alfa Aesar J61336.MC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies, Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, Synthesis of functionalized hydroxynaphthofurazan, Spectrophotometric and spectrofluorometric determination of clemastine hydrogen fumarate, loratadine, losartan potassium and ramipril in pharmaceutical formulations. Synthesis of 7-nitrobenzofurazan (NBD)-labeled maleimide, via Diels-Alder reaction. NBD-Cl is widely used to label peptides, proteins, drugs and other biomolecules. It is also a popular derivatizing reagent for HPLC analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-7-nitrobenzofurazan (NBD-chloride) was used in the following studies, Synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, Synthesis of functionalized hydroxynaphthofurazan, Spectrophoto
TRUSTED_SUSTAINABILITY

Specifications

CAS 29270-56-2
Melting Point 53°C to 57°C
Color Yellow
Flash Point 110°C (230°F)
Molecular Formula C6H2FN3O3
MDL Number MFCD00010196
Quantity 100 mg
Beilstein 1077571
Synonym 4-fluoro-7-nitrobenzofurazan, nbd-f, 4-fluoro-7-nitrobenzo c 1,2,5 oxadiazole, 7-fluoro-4-nitrobenzofurazan, 7-fluoro-4-nitrobenzo-2-oxa-1,3-diazole, 4-fluoro-7-nitrobenz-2-oxa-1,3-diazole, 2,1,3-benzoxadiazole, 4-fluoro-7-nitro, nbd-fluoride, nbd-f cpd, 4-fluoro-7-nitrobenzo c 1,2,5-oxadiazole
Solubility Information It is soluble in DMSO,DMF,CH3OH,CH3CN and CHCl3.
InChI Key PGZIDERTDJHJFY-UHFFFAOYSA-N
SMILES C1=C(C2=NON=C2C(=C1)F)[N+](=O)[O-]
IUPAC Name 4-fluoro-7-nitro-2,1,3-benzoxadiazole
Molecular Weight (g/mol) 183.098
PubChem CID 122123
Formula Weight 183.1
Physical Form Powder
Chemical Name or Material 4-Fluoro-7-nitrobenzofurazan
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS29J007 Certificate TypeCertificate of Analysis Date14/07/2023 Catalog NumberJ61336.MC
Lot NumberS20J009 Certificate TypeCertificate of Analysis Date27/06/2023 Catalog NumberJ61336.MC,J61336
Lot NumberW21I015 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog NumberJ61336.MC
Lot NumberX04I032 Certificate TypeCertificate of Analysis Date26/10/2022 Catalog NumberJ61336.MC
Lot NumberN09E028 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberJ61336.MC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-7-nitrobenzofurazan, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.