missing translation for 'onlineSavingsMsg'
Learn More
4-Fluoro-DL-phenylalanine, 98+%, Thermo Scientific Chemicals Product Code.: 11321967

4-Fluoro-DL-phenylalanine, 98+%, Thermo Scientific Chemicals

Product Code. 11321967
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11321967

Brand: Thermo Scientific Alfa Aesar L07585.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-65-0
Molecular Formula C9H10FNO2
Molecular Weight (g/mol) 183.18
MDL Number MFCD00002600,MFCD00063064
InChI Key XWHHYOYVRVGJJY-UHFFFAOYNA-N
Synonym 4-fluoro-dl-phenylalanine, 4-fluorophenylalanine, p-fluorophenylalanine, 2-amino-3-4-fluorophenyl propanoic acid, dl-p-fluorophenylalanine, p-fluoro-dl-phenylalanine, alnasid, dl-4-fluorophenylalanine, dl-4-f-phe-oh, h-dl-phe 4-f-oh
PubChem CID 4654
ChEBI CHEBI:84060
IUPAC Name 2-amino-3-(4-fluorophenyl)propanoic acid
SMILES NC(CC1=CC=C(F)C=C1)C(O)=O

Specifications

Melting Point ∽254°C (decomposition)
Beilstein 29338793
Quantity 5 g
Formula Weight 183.18
Percent Purity ≥98%
Chemical Name or Material 4-Fluoro-DL-phenylalanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10183455 Certificate TypeCertificate of Analysis Date07/09/2023 Catalog NumberL07585.06,L07585.03
Lot NumberS14J004 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog NumberL07585.06,L07585.03
Lot Number61000167 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL07585.06,L07585.03
Lot Number10183455 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL07585.06,L07585.03
Lot Number61080005 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL07585.06,L07585.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluoro-DL-phenylalanine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.