missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-fluorobenzyl)aniline, 97%, Thermo Scientific™

195.00 € - 599.00 €

Chemical Identifiers

CAS 80143-73-3
Molecular Formula C13H11F2N
Molecular Weight (g/mol) 219.235
MDL Number MFCD03210766
InChI Key OQVIJNAAQBTAAY-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzyl-4-fluorophenyl-amine
PubChem CID 1133337
IUPAC Name 4-fluoro-N-[(4-fluorophenyl)methyl]aniline
SMILES C1=CC(=CC=C1CNC2=CC=C(C=C2)F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472648
View Documents
Thermo Scientific Alfa Aesar
H56208.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15462648
View Documents
Thermo Scientific Alfa Aesar
H56208.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications

Chemical Identifiers

80143-73-3
219.235
OQVIJNAAQBTAAY-UHFFFAOYSA-N
1133337
C1=CC(=CC=C1CNC2=CC=C(C=C2)F)F
C13H11F2N
MFCD03210766
4-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzyl-4-fluorophenyl-amine
4-fluoro-N-[(4-fluorophenyl)methyl]aniline

Specifications

80143-73-3
MFCD03210766
OQVIJNAAQBTAAY-UHFFFAOYSA-N
4-fluoro-N-[(4-fluorophenyl)methyl]aniline
1133337
97%
C13H11F2N
4-fluoro-n-4-fluorophenyl methyl aniline, 4-fluorobenzyl-4-fluorophenyl-amine
C1=CC(=CC=C1CNC2=CC=C(C=C2)F)F
219.235
219.23
4-Fluoro-N-(4-fluorobenzyl)aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.