missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-N-(4-methoxybenzyl)aniline, 97%, Thermo Scientific™

72.50 € - 227.00 €

Chemical Identifiers

CAS 80143-71-1
Molecular Formula C14H14FNO
Molecular Weight (g/mol) 231.27
MDL Number MFCD00017946
InChI Key GSIXDRVRPADHPJ-UHFFFAOYSA-N
Synonym 4-fluoro-n-4-methoxybenzyl aniline, 4-fluoro-n-4-methoxyphenyl methyl aniline, n-4-methoxybenzyl-4-fluoroaniline, n-4-fluorophenyl-4-methoxybenzylamine
PubChem CID 4590297
IUPAC Name 4-fluoro-N-[(4-methoxyphenyl)methyl]aniline
SMILES COC1=CC=C(C=C1)CNC2=CC=C(C=C2)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472058
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55731.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15462058
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H55731.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

80143-71-1
231.27
GSIXDRVRPADHPJ-UHFFFAOYSA-N
4590297
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)F
C14H14FNO
MFCD00017946
4-fluoro-n-4-methoxybenzyl aniline, 4-fluoro-n-4-methoxyphenyl methyl aniline, n-4-methoxybenzyl-4-fluoroaniline, n-4-fluorophenyl-4-methoxybenzylamine
4-fluoro-N-[(4-methoxyphenyl)methyl]aniline

Specifications

80143-71-1
MFCD00017946
GSIXDRVRPADHPJ-UHFFFAOYSA-N
4-fluoro-N-[(4-methoxyphenyl)methyl]aniline
4590297
97%
C14H14FNO
4-fluoro-n-4-methoxybenzyl aniline, 4-fluoro-n-4-methoxyphenyl methyl aniline, n-4-methoxybenzyl-4-fluoroaniline, n-4-fluorophenyl-4-methoxybenzylamine
COC1=CC=C(C=C1)CNC2=CC=C(C=C2)F
231.27
231.27
4-Fluoro-N-(4-methoxybenzyl)aniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications