Learn More
4-Fluoro-N-methylaniline, 97%, Thermo Scientific Chemicals
39.10 € - 562.00 €
Chemical Identifiers
CAS | 459-59-6 |
---|---|
Molecular Formula | C7H8FN |
Molecular Weight (g/mol) | 125.146 |
MDL Number | MFCD00017942 |
InChI Key | VLWRKVBQUANIGI-UHFFFAOYSA-N |
PubChem CID | 68024 |
IUPAC Name | 4-fluoro-N-methylaniline |
SMILES | CNC1=CC=C(C=C1)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15411258
|
Thermo Scientific Alfa Aesar
H55105.03 |
1 g |
39.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421258
|
Thermo Scientific Alfa Aesar
H55105.06 |
5 g |
125.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15431258
|
Thermo Scientific Alfa Aesar
H55105.14 |
25 g | N/A | ||||||
Description
4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Fluoro-N-methylaniline is used as a model compound to study the biotransformation of secondary aromatic amines.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents , acids, acid chloride, acid anhydride and chloroformates.
Chemical Identifiers
459-59-6 | |
125.146 | |
VLWRKVBQUANIGI-UHFFFAOYSA-N | |
4-fluoro-N-methylaniline |
C7H8FN | |
MFCD00017942 | |
68024 | |
CNC1=CC=C(C=C1)F |
Specifications
459-59-6 | |
79°C (11 mmHg) | |
C7H8FN | |
MFCD00017942 | |
benzenamine, 4-fluoro-n-methyl, unii-88f75yis0f, n-methyl-4-fluoroaniline, acmc-1aem5, 4-fluoro-n-methyl aniline, 4-fluoro-n-methyl-aniline, 4-fluoro-n-methylbenzenamine, 4-fluorophenyl-methylamine, benzenamine,4-fluoro-n-methyl | |
CNC1=CC=C(C=C1)F | |
125.146 | |
125.15 | |
4-Fluoro-N-methylaniline |
1.04 | |
76°C (169°F) | |
1.532 | |
UN2810 | |
VLWRKVBQUANIGI-UHFFFAOYSA-N | |
4-fluoro-N-methylaniline | |
68024 | |
97% |
Safety and Handling
GHS H Statement
H318-H302-H317-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 207-294-1
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only