missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylmagnesium chloride, 0.25M solution in THF, AcroSeal™

Product Code. 10133194 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10133194

Brand: Thermo Scientific Acros 426790500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

CAS: 1643-73-8 | C7H6ClFMg | 168.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Especificaciones

Color Brown or Orange to Yellow
CAS 1643-73-8
CAS Min % 2.67
CAS Max % 6.24
Molecular Formula C7H6ClFMg
Linear Formula FC6H4CH2MgCl
MDL Number MFCD01319898
Concentration 0.15 to 0.35M (exact strength on the certificate of analysis)
Density 0.9480g/mL
Synonym 4-fluorobenzylmagnesium chloride, chloro 4-fluorophenyl methyl magnesium, 4-fluorobenzylmagnesiumchloride, 4-fluorobenzyl magnesium chloride, pqdgqupddguklp-uhfffaoysa-m, 4-fluoro benzylmagnesium chloride, 4-fluorobenzylmagnesium chloride, 0.25 m in thf, 4-fluorobenzylmagnesium chloride 0.25 m in tetrahydrofuran, 4-fluorobenzylmagnesium chloride, 0.25m w/w in thf
InChI Key PQDGQUPDDGUKLP-UHFFFAOYSA-M
SMILES FC1=CC=C(C[Mg]Cl)C=C1
IUPAC Name magnesium;1-fluoro-4-methanidylbenzene;chloride
Molecular Weight (g/mol) 168.88
PubChem CID 2778244
Formula Weight 168.88
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.948
Quantity 50 mL
Physical Form Solution
Chemical Name or Material 4-Fluorobenzylmagnesium chloride, 0.25M solution in THF
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • 4-Fluorobenzylmagnesium chloride, 0.25M solution in THF
Palabra de advertencia
  • Danger
Clasificación
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado