Learn More
4-Fluorophenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals
103.00 € - 514.00 €
Chemical Identifiers
CAS | 352-13-6, 60-29-7 |
---|---|
Molecular Formula | C6H4BrFMg |
Molecular Weight (g/mol) | 199.31 |
MDL Number | MFCD00009667 |
InChI Key | QYBFFRXNNFXREA-UHFFFAOYSA-M |
Synonym | 4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent |
PubChem CID | 2734897 |
SMILES | FC1=CC=C([Mg]Br)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10402242
|
Thermo Scientific Acros
377371000 |
100 mL | AcroSeal Glass Bottle in Tin Can |
103.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10597303
|
Thermo Scientific
377378000 |
800 mL | AcroSeal™ Glass Bottle |
514.00 €
800mL Qty Price Break Available |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
352-13-6, 60-29-7 | |
199.31 | |
QYBFFRXNNFXREA-UHFFFAOYSA-M | |
2734897 |
C6H4BrFMg | |
MFCD00009667 | |
4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent | |
FC1=CC=C([Mg]Br)C=C1 |
Specifications
Undesignated | |
38.22, 57.96 | |
C6H4BrFMg | |
MFCD00009667 | |
1.0430g/mL | |
QYBFFRXNNFXREA-UHFFFAOYSA-M | |
bromo(4-fluorophenyl)magnesium | |
2734897 | |
Solution |
352-13-6, 60-29-7 | |
42.04, 61.78 | |
F(C6H4)MgBr | |
2 to 2.2M | |
4-fluorophenylmagnesium bromide, magnesium, bromo 4-fluorophenyl, bromo 4-fluorophenyl magnesium, p-fluorophenylmagnesium bromide, 4-fluorophenyl magnesium bromide, 4-f-c6h4-mgbr, 4-fluorophenylmagnesiumbromide, 4-fluorphenylmagnesium bromide, 4-flourophenylmagnesium bromide, 4-fluorophenyl magnesiumbromide, grignard reagent | |
FC1=CC=C([Mg]Br)C=C1 | |
199.31 | |
199.3 | |
4-Fluorophenylmagnesium bromide, 2M Solution in Diethyl Ether |
Safety and Handling
GHS P Statement Harmful if swallowed. May cause drowsiness or dizziness. Causes severe skin burns and eye damage. In contact with water releases flammable gas. Extremely flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking. Reacts violently with water.
GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.