missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoropropiophenone, 97+%, Thermo Scientific Chemicals

Product Code. 11463317
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 456-03-1
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.168
MDL Number MFCD00000356
InChI Key QIJNVLLXIIPXQT-UHFFFAOYSA-N
Synonym 4'-fluoropropiophenone, 1-4-fluorophenyl propan-1-one, p-fluoropropiophenone, 4-fluoropropiophenone, ethyl 4-fluorophenyl ketone, 1-propanone, 1-4-fluorophenyl, propiophenone, 4'-fluoro, para-fluoropropiophenone, 1-4-fluorophenyl-1-propanone, 4;-fluoropropiophenone
PubChem CID 68004
IUPAC Name 1-(4-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(C=C1)F

Specifications

Density 1.096
Boiling Point 100°C to 102°C (22 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.5059
Quantity 10 g
Beilstein 1210310
Formula Weight 152.17
Percent Purity ≥97%
Chemical Name or Material 4'-Fluoropropiophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'