missing translation for 'onlineSavingsMsg'
Learn More
4-Fluorothiophenol, 97%, Thermo Scientific Chemicals Product Code.: 11428227

4-Fluorothiophenol, 97%, Thermo Scientific Chemicals

Product Code. 11428227
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11428227

Brand: Thermo Scientific Alfa Aesar A17600.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the synthesis of Bicalutamide, an anti-cancer drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of Bicalutamide, an anti-cancer drug.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents and air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-42-6
Molecular Formula C6H5FS
Molecular Weight (g/mol) 128.164
MDL Number MFCD00004846
InChI Key OKIHXNKYYGUVTE-UHFFFAOYSA-N
Synonym 4-fluorothiophenol, p-fluorothiophenol, 4-fluorobenzene-1-thiol, 4-fluoro thiophenol, p-fluorophenyl mercaptan, benzenethiol, 4-fluoro, 4-fluoro-benzenethiol, 4-mercaptofluorobenzene, 4-fluorthiophenol, p-fluoro-thiophenol
PubChem CID 67789
IUPAC Name 4-fluorobenzenethiol
SMILES C1=CC(=CC=C1F)S

Specifications

Density 1.203
Boiling Point 170°C to 172°C
Flash Point 57°C (135°F)
Odor Unpleasant
Refractive Index 1.55
Quantity 5 g
UN Number UN3071
Beilstein 1446350
Sensitivity Air Sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 128.17
Percent Purity ≥98%
Assay (dry wt.), water <7%
Chemical Name or Material 4-Fluorothiophenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242176 Certificate TypeCertificate of Analysis Date27/07/2022 Catalog NumberA17600.14,A17600.06,A17600.22
Lot Number10141841 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17600.14,A17600.06,A17600.22
Lot Number10113504 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17600.14,A17600.06,A17600.22
Lot Number10161696 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17600.14,A17600.06,A17600.22
Lot Number10157873 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17600.14,A17600.06,A17600.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Fluorothiophenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.