missing translation for 'onlineSavingsMsg'
Learn More

4-Hexylresorcinol, 98%, Thermo Scientific Chemicals

Product Code. 10674702
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-77-6
Molecular Formula C12H18O2
Molecular Weight (g/mol) 194.27
MDL Number MFCD00002284
InChI Key WFJIVOKAWHGMBH-UHFFFAOYSA-N
Synonym hexylresorcinol, 4-hexylresorcinol, antascarin, ascaricid, ascarinol, 4-n-hexylresorcinol, oxana, p-hexylresorcinol, adrover, caprokol
PubChem CID 3610
IUPAC Name 4-hexylbenzene-1,3-diol
SMILES CCCCCCC1=CC=C(O)C=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 65.0°C to 70.0°C
Color Orange to White or Pink
Boiling Point 333.0°C to 335.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 06, II, 904
Merck Index 15, 4748
Solubility Information Solubility in water: 0.5g/l (18°C). Other solubilities: soluble in alcohol,glycerol,ether,chloroform,,acetone and vegetable oils
Formula Weight 194.27
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Hexylresorcinol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'