missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3,5-diiodobenzonitrile, 97%, Thermo Scientific Chemicals

77.00 € - 910.00 €

Chemical Identifiers

CAS 1689-83-4
Molecular Formula C7H3I2NO
Molecular Weight (g/mol) 370.92
MDL Number MFCD00016422
InChI Key NRXQIUSYPAHGNM-UHFFFAOYSA-N
Synonym ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox
PubChem CID 15530
ChEBI CHEBI:81821
IUPAC Name 4-hydroxy-3,5-diiodobenzonitrile
SMILES OC1=C(I)C=C(C=C1I)C#N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11401658
View Documents
Thermo Scientific Alfa Aesar
A13929.06
5 g
77.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11411658
View Documents
Thermo Scientific Alfa Aesar
A13929.14
25 g
297.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
11421658
View Documents
Thermo Scientific Alfa Aesar
A13929.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1689-83-4
370.92
NRXQIUSYPAHGNM-UHFFFAOYSA-N
15530
4-hydroxy-3,5-diiodobenzonitrile
C7H3I2NO
MFCD00016422
ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox
CHEBI:81821
OC1=C(I)C=C(C=C1I)C#N

Specifications

1689-83-4
Odorless
MFCD00016422
2364041
NRXQIUSYPAHGNM-UHFFFAOYSA-N
4-hydroxy-3,5-diiodobenzonitrile
15530
370.92
4-Hydroxy-3,5-diiodobenzonitrile
200°C to 207°C
C7H3I2NO
UN3439
ioxynil, actril, bentrol, 3,5-diiodo-4-hydroxybenzonitrile, cipotril, trevespan, certrol, joxynil, loxynil, iotox
OC1=C(I)C=C(C=C1I)C#N
370.92
CHEBI:81821
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H361-H373-H312-H319
Toxic if swallowed.
Toxic if inhaled.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement
P260-P261-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

missing translation for 'rtecsNumber' : DI4025000

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.