missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxy-3,5-dimethoxycinnamic acid, 98%, Thermo Scientific Chemicals Product Code.: 11379519

4-Hydroxy-3,5-dimethoxycinnamic acid, 98%, Thermo Scientific Chemicals

Product Code. 11379519
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11379519

Brand: Thermo Scientific Alfa Aesar A15676.03

This item is not returnable. View return policy

4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxy-3,5-dimethoxycinnamic acid is a commonly used matrix in MALDI mass spectrometry. It is a useful matrix for a wide variety of peptides and proteins. It serves well as a matrix for MALDI due to its ability to absorb laser radiation and to also donate protons (H+) to the analyte of interest.

Solubility
Soluble in methanol, ethanol and dimethyl sulfoxide.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-59-6
Molecular Formula C11H12O5
Molecular Weight (g/mol) 224.212
MDL Number MFCD00004401
InChI Key PCMORTLOPMLEFB-ONEGZZNKSA-N
Synonym sinapic acid, sinapinic acid, 3,5-dimethoxy-4-hydroxycinnamic acid, sinapinate, sinapate, trans-sinapic acid, 4-hydroxy-3,5-dimethoxycinnamic acid, synapoic acid, 2e-3-4-hydroxy-3,5-dimethoxyphenyl prop-2-enoic acid, unii-p0i60993ec
PubChem CID 637775
ChEBI CHEBI:15714
IUPAC Name (E)-3-(4-hydroxy-3,5-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1O)OC)C=CC(=O)O

Specifications

Melting Point ∼195°C (decomposition)
Quantity 1 g
Beilstein 2130006
Solubility Information Soluble in methanol,ethanol and dimethyl sulfoxide.
Formula Weight 224.21
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxycinnamic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10213964 Certificate TypeCertificate of Analysis Date31/05/2023 Catalog NumberA15676.03
Lot Number10238633 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA15676.03,A15676.14,A15676.06
Lot Number10165350 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15676.03,A15676.14,A15676.06
Lot NumberJ9878A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15676.03,A15676.14,A15676.06
Lot Number10213964 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15676.03,A15676.14,A15676.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxy-3,5-dimethoxycinnamic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.