Learn More
4'-Hydroxy-3'-methoxyacetophenone, 98%, Thermo Scientific Chemicals
48.34 € - 200.00 €
Chemical Identifiers
CAS | 498-02-2 |
---|---|
Molecular Formula | C9H10O3 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00008747 |
InChI Key | DFYRUELUNQRZTB-UHFFFAOYSA-N |
Synonym | acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon |
PubChem CID | 2214 |
ChEBI | CHEBI:2781 |
IUPAC Name | 1-(4-hydroxy-3-methoxyphenyl)ethanone |
SMILES | COC1=CC(=CC=C1O)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448653
|
Thermo Scientific Alfa Aesar
A10439.18 |
50 g |
48.34 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458653
|
Thermo Scientific Alfa Aesar
A10439.30 |
250 g |
124.62 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344719
|
Thermo Scientific Alfa Aesar
A10439.36 |
500 g |
200.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4'-Hydroxy-3'-methoxyacetophenone is used as an inhibitor of nicotinamide adenine dinucleotide phosphate (NADPH) oxidase, which is an enzyme responsible for reactive oxygen species production. It is also useful in the treatment of various inflammatory diseases and atherosclerosis. Further, it shows an anti-inflammatory activity and improves endothelial function by reducing oxidative stress. In addition to this, it is used for studying nitric oxide regulated processes in plants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in ethanol, chloroform, ether, hot water, benzene, ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
498-02-2 | |
166.18 | |
DFYRUELUNQRZTB-UHFFFAOYSA-N | |
2214 | |
1-(4-hydroxy-3-methoxyphenyl)ethanone |
C9H10O3 | |
MFCD00008747 | |
acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon | |
CHEBI:2781 | |
COC1=CC(=CC=C1O)C(C)=O |
Specifications
498-02-2 | |
295°C to 300°C | |
MFCD00008747 | |
14,741 | |
Soluble in ethanol,chloroform,ether,hot water,benzene,ethyl acetate and dimethyl sulfoxide. Insoluble in petroleum ether. | |
COC1=CC(=CC=C1O)C(C)=O | |
166.18 | |
CHEBI:2781 | |
98% |
112°C to 116°C | |
C9H10O3 | |
637373 | |
acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon | |
DFYRUELUNQRZTB-UHFFFAOYSA-N | |
1-(4-hydroxy-3-methoxyphenyl)ethanone | |
2214 | |
166.18 | |
4'-Hydroxy-3'-methoxyacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 207-854-5
missing translation for 'rtecsNumber' : AM8800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only