Learn More
4-Hydroxy-3-methoxycinnamic acid, 99%, Thermo Scientific Chemicals
29.30 € - 171.00 €
Chemical Identifiers
CAS | 1135-24-6 |
---|---|
Molecular Formula | C10H10O4 |
Molecular Weight (g/mol) | 194.19 |
MDL Number | MFCD00004400 |
InChI Key | KSEBMYQBYZTDHS-HWKANZROSA-N |
Synonym | ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl |
PubChem CID | 445858 |
ChEBI | CHEBI:17620 |
IUPAC Name | (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid |
SMILES | COC1=C(C=CC(=C1)C=CC(=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10367950
|
Thermo Scientific Acros
156360050 |
5 g | Glass bottle |
29.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637512
|
Thermo Scientific Acros
156360250 |
25 g | Glass bottle |
53.70 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10593511
|
Thermo Scientific Acros
156361000 |
100 g | Glass bottle |
171.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1135-24-6 | |
194.19 | |
KSEBMYQBYZTDHS-HWKANZROSA-N | |
445858 | |
(E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid |
C10H10O4 | |
MFCD00004400 | |
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl | |
CHEBI:17620 | |
COC1=C(C=CC(=C1)C=CC(=O)O)O |
Specifications
1135-24-6 | |
100.0 | |
White to Beige | |
98.5% min. (HPLC) | |
HOC6H3(OCH3)CH=CHCO2H | |
10, 436 | |
ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl | |
KSEBMYQBYZTDHS-HWKANZROSA-N | |
(E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid | |
445858 | |
194.19 | |
Powder |
98.5 | |
169.0°C to 173.0°C | |
Authentic | |
C10H10O4 | |
MFCD00004400 | |
15, 4089 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol and ethyl acetate,moderately soluble in diethyl ether,sparingly soluble in petroleum ether and benzene | |
COC1=C(C=CC(=C1)C=CC(=O)O)O | |
194.19 | |
CHEBI:17620 | |
99% | |
4-Hydroxy-3-methoxycinnamic acid, 99% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 214-490-0
missing translation for 'rtecsNumber' : UD3365500
missing translation for 'tsca' : TSCA
- 4-Hydroxy-3-methoxycinnamic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only