Learn More
4-Hydroxy-3-nitrophenylacetic acid, 99%, Thermo Scientific Chemicals
36.88 € - 430.14 €
Chemical Identifiers
CAS | 10463-20-4 |
---|---|
Molecular Formula | C8H7NO5 |
Molecular Weight (g/mol) | 197.146 |
MDL Number | MFCD00007122 |
InChI Key | QBHBHOSRLDPIHG-UHFFFAOYSA-N |
Synonym | 4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629 |
PubChem CID | 447364 |
ChEBI | CHEBI:546274 |
SMILES | C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11471518
|
Thermo Scientific Alfa Aesar
A13858.03 |
1 g | N/A | ||||||
11481518
|
Thermo Scientific Alfa Aesar
A13858.06 |
5 g |
128.64 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11491518
|
Thermo Scientific Alfa Aesar
A13858.14 |
25 g |
430.14 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10463-20-4 | |
197.146 | |
QBHBHOSRLDPIHG-UHFFFAOYSA-N | |
447364 | |
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O |
C8H7NO5 | |
MFCD00007122 | |
4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629 | |
CHEBI:546274 |
Specifications
10463-20-4 | |
C8H7NO5 | |
2696044 | |
QBHBHOSRLDPIHG-UHFFFAOYSA-N | |
197.146 | |
CHEBI:546274 | |
99% |
146°C to 148°C | |
MFCD00007122 | |
4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629 | |
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O | |
447364 | |
197.15 | |
4-Hydroxy-3-nitrophenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 233-948-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.