missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide, 98%, Thermo Scientific™ Product Code.: 11461167

4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide, 98%, Thermo Scientific™

Product Code. 11461167
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11461167

Brand: Thermo Scientific Alfa Aesar B24153.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54714-10-2
Molecular Formula C16H12O4S
Molecular Weight (g/mol) 300.328
MDL Number MFCD00075268
InChI Key UIVWXNPUCAHAJX-UHFFFAOYSA-N
Synonym 4-hydroxy-2,5-diphenylthiophen-3 2h-one 1,1-dioxide, 2,5-diphenyl-4-hydroxy-3-oxo-2,3-dihydrothiophene 1,1-dioxide, 4-hydroxy-2,5-diphenyl-3 2h-thiophenone 1,1-dioxide, 4-hydroxy-2,5-diphenyl-2-hydrothiophene-1,1,3-trione, 4-hydroxy-2,5-diphenyl-2h-1??-thiophene-1,1,3-trione, hotdo, acmc-1apg9, 2,5-diphenyl-4-hydroxy-3-thiophenone 1,1-dioxide, 4-hydroxy-1,1-dioxo-2,5-diphenyl-thiophen-3-one, 4-hydroxy-2,5-diphenyl-3-thiophenone 1,1-dioxideShow MoreShow Less
PubChem CID 388546
IUPAC Name 4-hydroxy-1,1-dioxo-2,5-diphenylthiophen-3-one
SMILES C1=CC=C(C=C1)C2C(=O)C(=C(S2(=O)=O)C3=CC=CC=C3)O

Specifications

Melting Point 238°C to 241°C
Quantity 25 g
Beilstein 544870
Formula Weight 300.34
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxy-3-oxo-2,5-diphenyl-2,3-dihydrothiophene 1,1-dioxide, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.