Learn More
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde, 98+%, Thermo Scientific Chemicals
65.80 € - 175.00 €
Chemical Identifiers
CAS | 53104-95-3 |
---|---|
Molecular Formula | C8H5F3O3 |
Molecular Weight (g/mol) | 206.12 |
MDL Number | MFCD04973780 |
InChI Key | GPJSLRQIOKFRFL-UHFFFAOYSA-N |
Synonym | 4-hydroxy-3-trifluoromethoxy benzaldehyde, benzaldehyde, 4-hydroxy-3-trifluoromethoxy, 4-hydroxy-3-trifluoromethoxy benzaldehyde, 98+%, 4-oxidanyl-3-trifluoromethyloxy benzaldehyde |
PubChem CID | 7018050 |
IUPAC Name | 4-hydroxy-3-(trifluoromethoxy)benzaldehyde |
SMILES | OC1=C(OC(F)(F)F)C=C(C=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468135
|
Thermo Scientific Alfa Aesar
H32602.MD |
250 mg |
65.80 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15405357
|
Thermo Scientific Alfa Aesar
H32602.03 |
1 g |
175.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hydroxy-3-(trifluoromethoxy)benzaldehyde is a fluorinated compound, which is used for biochemical research.
Solubility
Insoluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Chemical Identifiers
53104-95-3 | |
206.12 | |
GPJSLRQIOKFRFL-UHFFFAOYSA-N | |
7018050 | |
OC1=C(OC(F)(F)F)C=C(C=O)C=C1 |
C8H5F3O3 | |
MFCD04973780 | |
4-hydroxy-3-trifluoromethoxy benzaldehyde, benzaldehyde, 4-hydroxy-3-trifluoromethoxy, 4-hydroxy-3-trifluoromethoxy benzaldehyde, 98+%, 4-oxidanyl-3-trifluoromethyloxy benzaldehyde | |
4-hydroxy-3-(trifluoromethoxy)benzaldehyde |
Specifications
53104-95-3 | |
C8H5F3O3 | |
Air Sensitive | |
Insoluble in water. | |
OC1=C(OC(F)(F)F)C=C(C=O)C=C1 | |
206.12 | |
206.12 | |
4-Hydroxy-3-(trifluoromethoxy)benzaldehyde |
76°C to 80°C | |
MFCD04973780 | |
4-hydroxy-3-trifluoromethoxy benzaldehyde, benzaldehyde, 4-hydroxy-3-trifluoromethoxy, 4-hydroxy-3-trifluoromethoxy benzaldehyde, 98+%, 4-oxidanyl-3-trifluoromethyloxy benzaldehyde | |
GPJSLRQIOKFRFL-UHFFFAOYSA-N | |
4-hydroxy-3-(trifluoromethoxy)benzaldehyde | |
7018050 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.