missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-2-pyrone, 98%, Thermo Scientific Chemicals

Product Code. 11317786
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-10-5
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006641,MFCD18820248
InChI Key OOKCZXGEYPSNIM-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-2-pyrone, 2-hydroxy-6-methyl-4h-pyran-4-one, 4-hydroxy-6-methyl-2h-pyran-2-one, triacetic acid lactone, triacetate lactone, 2h-pyran-2-one, 4-hydroxy-6-methyl, 6-methyl-4-hydroxypyron-2, unii-s1s883s4ee, 6-methyl-4-hydroxy-2-pyrone, ccris 3600
PubChem CID 54675757
ChEBI CHEBI:16458
IUPAC Name 4-hydroxy-6-methylpyran-2-one
SMILES CC1=CC(=O)C=C(O)O1

Specifications

Melting Point ∽186°C (decomposition)
Quantity 25 g
Beilstein 113815
Formula Weight 126.11
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-6-methyl-2-pyrone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'