missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzaldehyde, 99%, Thermo Scientific Chemicals

Product Code. 10184460
Click to view available options
Quantity:
50 g
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-08-0
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
InChI Key RGHHSNMVTDWUBI-UHFFFAOYSA-N
Synonym p-hydroxybenzaldehyde, 4-formylphenol, p-formylphenol, p-oxybenzaldehyde, benzaldehyde, 4-hydroxy, parahydroxybenzaldehyde, benzaldehyde, p-hydroxy, 4-hydroxy-benzaldehyde, usaf m-6, 4-hydroxy benzaldehyde
PubChem CID 126
ChEBI CHEBI:17597
IUPAC Name 4-hydroxybenzaldehyde
SMILES C1=CC(=CC=C1C=O)O

Specifications

Melting Point 114.0°C to 118.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H4CHO
Quantity 250 g
Beilstein 08,64
Merck Index 15,4849
Solubility Information Solubility in water: 13g/L (30°C). Other solubilities: 36.8g/L benzene (65°C)
Formula Weight 122.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'