missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

63.80 € - 222.00 €

Chemical Identifiers

CAS 269409-70-3
Molecular Formula C12H17BO3
Molecular Weight (g/mol) 220.075
MDL Number MFCD02093756
InChI Key BICZJRAGTCRORZ-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
PubChem CID 2734624
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11387419
View Documents
Thermo Scientific Alfa Aesar
H27836.03
1 g
63.80 €
1g
Please to purchase this item. Need a web account? Register with us today!
11397419
View Documents
Thermo Scientific Alfa Aesar
H27836.06
5 g
222.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

269409-70-3
220.075
BICZJRAGTCRORZ-UHFFFAOYSA-N
2734624
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
C12H17BO3
MFCD02093756
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Specifications

269409-70-3
C12H17BO3
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacol ester, 4-hydroxyphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenol, 4-hydroxyphenylboronic acid pinacolate, 2-4-hydroxyphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-hydroxyphenylboronicacidpinacolester, 4-hydroxybenzeneboronic acid pinacol ester, phenol, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-hydroxyphenylboronic acid pinacol cyclic ester
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)O
220.075
220.07
4-Hydroxybenzeneboronic acid pinacol ester
114°C to 117°C
MFCD02093756
BICZJRAGTCRORZ-UHFFFAOYSA-N
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
2734624
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.