Learn More
4-Hydroxybenzoic Acid, 99+%, Thermo Scientific Chemicals
25.02 € - 72.16 €
Chemical Identifiers
CAS | 99-96-7 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00002547 |
InChI Key | FJKROLUGYXJWQN-UHFFFAOYSA-N |
Synonym | p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german |
PubChem CID | 135 |
ChEBI | CHEBI:30763 |
IUPAC Name | 4-hydroxybenzoic acid |
SMILES | OC(=O)C1=CC=C(O)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10676882
|
Thermo Scientific Acros
120991000 |
100 g | Plastic bottle |
25.02 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10754051
|
Thermo Scientific Acros
120995000 |
500 g | Plastic bottle |
57.49 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10060970
|
Thermo Scientific Acros
120990010 |
1 kg | Plastic bottle |
72.16 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
99-96-7 | |
138.12 | |
FJKROLUGYXJWQN-UHFFFAOYSA-N | |
135 | |
4-hydroxybenzoic acid |
C7H6O3 | |
MFCD00002547 | |
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german | |
CHEBI:30763 | |
OC(=O)C1=CC=C(O)C=C1 |
Specifications
99-96-7 | |
100.0 | |
Ivory to White | |
Authentic | |
C7H6O3 | |
MFCD00002547 | |
15, 4850 | |
Solubility in water: 5g/L (20°C). Other solubilities: soluble in alcohol and ether,slightly soluble in chloroform,practically insoluble in carbon disulfide | |
OC(=O)C1=CC=C(O)C=C1 | |
138.12 | |
CHEBI:30763 | |
99+% | |
4-Hydroxybenzoic acid, 99% |
99.0 | |
214°C to 217°C | |
199°C | |
99% min. (HPLC) | |
HOC6H4CO2H | |
10, 149 | |
p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german | |
FJKROLUGYXJWQN-UHFFFAOYSA-N | |
4-hydroxybenzoic acid | |
135 | |
138.12 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-804-9
missing translation for 'rtecsNumber' : DH1925000
missing translation for 'tsca' : TSCA
- 4-Hydroxybenzoic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
RUO – Research Use Only