missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxybenzylideneacetone, 97%, Thermo Scientific Chemicals Product Code.: 11408237

4-Hydroxybenzylideneacetone, 97%, Thermo Scientific Chemicals

Product Code. 11408237
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11408237

Brand: Thermo Scientific Alfa Aesar A17605.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Hydroxybenzylideneacetone is used in the synthesis of 4-(4-hydroxy-phenyl)-butan-2-one. This reaction needs the reagents of cyclohexane and AlCl3, and the solvent of CH2Cl2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzylideneacetone is used in the synthesis of 4-(4-hydroxy-phenyl)-butan-2-one. This reaction needs the reagents of cyclohexane and AlCl3, and the solvent of CH2Cl2.

Solubility
Optimally soluble in aqueous solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3160-35-8
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00016490
InChI Key OCNIKEFATSKIBE-NSCUHMNNSA-N
Synonym p-hydroxybenzalacetone, 4-hydroxybenzylideneacetone, 4-hydroxybenzal acetone, 4-hydroxycinnamoylmethane, 4-hydroxybenzalacetone, p-hydroxybenzylidene acetone, 4-p-hydroxyphenyl-3-buten-2-one, 4-4-hydroxyphenyl but-3-en-2-one, 3-buten-2-one, 4-4-hydroxyphenyl, 3-buten-2-one, 4-p-hydroxyphenyl
PubChem CID 796857
SMILES CC(=O)\C=C\C1=CC=C(O)C=C1

Specifications

Melting Point 106°C to 111°C
Quantity 25 g
Beilstein 2412544
Solubility Information Optimally soluble in aqueous solvents.
IUPAC Name (E)-4-(4-hydroxyphenyl)but-3-en-2-one
Formula Weight 162.19
Percent Purity 97%
Chemical Name or Material 4-Hydroxybenzylideneacetone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxybenzylideneacetone, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.