missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxychalcone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents.
4-Hydroxychalcone is used as anti-angiogenesis, antineoplastic. It is also used as organic intermediates.
Solubility
Insoluble in water.
Notes
Store at 4°C. Incompatible with oxidizing agents.
Specifications
Specifications
Melting Point | 183°C to 185°C |
Quantity | 25 g |
Beilstein | 2049155 |
Solubility Information | Insoluble in water. |
Formula Weight | 224.26 |
Percent Purity | 97% |
Chemical Name or Material | 4-Hydroxychalcone |
Safety and Handling
Product Identifier
- 4-Hydroxychalcone
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 4-Hydroxychalcone
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction