missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycoumarin, 98+%, Thermo Scientific Chemicals

Product Code. 11496997
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Hydroxycoumarin is involved in annulation reactions, due to the relatively high acidity of the C-H bond at the 3-position: a three-component reaction with isocyanides and dialkyl acetylene dicarboxylates affords annulated 4H-pyrans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxycoumarin is involved in annulation reactions, due to the relatively high acidity of the C-H bond at the 3-position: a three-component reaction with isocyanides and dialkyl acetylene dicarboxylates affords annulated 4H-pyrans.

Solubility
Soluble in ethanol and dimethyl formamide. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1076-38-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006856
InChI Key OWBBAPRUYLEWRR-UHFFFAOYSA-N
Synonym 4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
PubChem CID 54682930
ChEBI CHEBI:40070
IUPAC Name 4-hydroxychromen-2-one
SMILES OC1=CC(=O)C2=CC=CC=C2O1

Specifications

Melting Point ∼213°C (decomposition)
Odor Odorless
Quantity 100 g
Beilstein 129768
Solubility Information Soluble in ethanol and dimethyl formamide. Insoluble in water.
Formula Weight 162.14
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxycoumarin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'