missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylacetic acid, 99%, Thermo Scientific Chemicals

Product Code. 11407267
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a reagent in the acylation of phenols and amines. It is also used for fluorometric determination of oxidative enzymes.

Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-38-7
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004347
InChI Key XQXPVVBIMDBYFF-UHFFFAOYSA-N
Synonym 4-hydroxyphenylacetic acid, 2-4-hydroxyphenyl acetic acid, p-hydroxyphenylacetic acid, 4-hydroxyphenyl acetic acid, 4-hydroxyphenylacetate, 4-hydroxybenzeneacetic acid, benzeneacetic acid, 4-hydroxy, p-hydroxyphenyl acetic acid, parahydroxy phenylacetic acid, 4-carboxymethylphenol
PubChem CID 127
ChEBI CHEBI:18101
IUPAC Name 2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1CC(=O)O)O

Specifications

Melting Point 148°C to 150°C
Quantity 100 g
Beilstein 1448766
Solubility Information Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 4-Hydroxyphenylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.