missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10522242
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71597-85-8
Molecular Formula C6H7BO3
Molecular Weight (g/mol) 137.93
MDL Number MFCD01074628
InChI Key COIQUVGFTILYGA-UHFFFAOYSA-N
Synonym 4-hydroxyphenyl boronic acid, 4-hydroxybenzeneboronic acid, 4-boronophenol, 4-hydroxyphenyl boranediol, p-hydroxyphenylboronic acid, 4-hydroxyphenyl-boronic acid, boronic acid, 4-hydroxyphenyl, pubchem1729, acmc-209ojb
PubChem CID 2734360
IUPAC Name (4-hydroxyphenyl)boronic acid
SMILES OB(O)C1=CC=C(O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 285.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: 25g/L
Formula Weight 137.93
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Hydroxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'