missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxypiperidine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 86°C to 90°C |
pH | 11.5 to 11.7 |
Boiling Point | 108°C to 114°C (10 mmHg) |
Flash Point | 107°C (224°F) |
Odor | Amine-like |
Quantity | 5 g |
UN Number | UN3263 |
Beilstein | 102738 |
Sensitivity | Hygroscopic |
Formula Weight | 101.15 |
Show More |
Safety and Handling
Product Identifier
- 4-Hydroxypiperidine
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Piperidin-4-ol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'