missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11475867
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as intermediate in organic syntheses & in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as intermediate in organic syntheses & in medicine.

Solubility
Reacts with water.

Notes
Moisture & light sensitive. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-61-3
Molecular Formula C6H4ClIO2S
Molecular Weight (g/mol) 302.51
MDL Number MFCD00007441
InChI Key POXFXTSTVWDWIR-UHFFFAOYSA-N
Synonym pipsyl chloride, 4-iodobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-iodo, p-iodobenzenesulfonyl chloride, 4-iodobenzenesulphonyl chloride, 4-iodobenzenesulfonylchloride, 4-iodophenylsulfonyl chloride, iodobenzene-p-sulfonyl chloride, unii-mvl274ezog, mvl274ezog
PubChem CID 7399
IUPAC Name 4-iodobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)I

Specifications

Melting Point 79°C to 84°C
Quantity 5 g
UN Number UN3261
Beilstein 2691661
Sensitivity Light and moisture sensitive
Merck Index 14,7486
Solubility Information Reacts with water.
Formula Weight 302.52
Percent Purity 97%
Chemical Name or Material 4-Iodobenzenesulfonyl chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'