missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenol, 99%, Thermo Scientific Chemicals

53.97 € - 1111.00 €

Chemical Identifiers

CAS 540-38-5
Molecular Formula C6H5IO
Molecular Weight (g/mol) 220.01
MDL Number MFCD00002327
InChI Key VSMDINRNYYEDRN-UHFFFAOYSA-N
Synonym p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
PubChem CID 10894
ChEBI CHEBI:43521
IUPAC Name 4-iodophenol
SMILES OC1=CC=C(I)C=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10153260
View Documents
Thermo Scientific Acros
122390100
10 g Glass bottle
53.97 €
10g
Please to purchase this item. Need a web account? Register with us today!
10265260
View Documents
Thermo Scientific Acros
122390250
25 g Glass bottle
91.15 €
25g
Please to purchase this item. Need a web account? Register with us today!
11944681
View Documents
Thermo Scientific Acros
122391000
100 g Glass bottle
266.33 €
100g
Please to purchase this item. Need a web account? Register with us today!
15129173
View Documents
Thermo Scientific Acros
122395000
500 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

540-38-5
220.01
VSMDINRNYYEDRN-UHFFFAOYSA-N
10894
4-iodophenol
C6H5IO
MFCD00002327
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
CHEBI:43521
OC1=CC=C(I)C=C1

Specifications

540-38-5
Beige to Brown or Pink
Authentic
C6H5IO
MFCD00002327
15,5079
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and ether
OC1=CC=C(I)C=C1
220.01
CHEBI:43521
99%
4-Iodophenol
89.0°C to 94.0°C
287.0°C
98.5% min. (GC)
IC6H4OH
06,208
p-iodophenol, phenol, 4-iodo, phenol, p-iodo, 4-hydroxyiodobenzene, p-hydroxyiodobenzene, 4-hydroxyphenyl iodide, 4-iodo-phenol, unii-bh194bak0b, ccris 668, bh194bak0b
VSMDINRNYYEDRN-UHFFFAOYSA-N
4-iodophenol
10894
220.01
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 208-745-5

Product Identifier
  • 4-Iodophenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.