missing translation for 'onlineSavingsMsg'
Learn More

4-Iodotoluene, 98%, Thermo Scientific Chemicals

Product Code. 10607492
Click to view available options
Quantity:
10 g
50 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-31-7
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001059
InChI Key UDHAWRUAECEBHC-UHFFFAOYSA-N
Synonym 4-iodotoluene, p-iodotoluene, benzene, 1-iodo-4-methyl, toluene, p-iodo, p-tolyl iodide, p-methyliodobenzene, 1-methyl-4-iodobenzene, 1-iodo-4-methyl-benzene, unii-g75kcn1am8, iodotoluene 4-
PubChem CID 12207
IUPAC Name 1-iodo-4-methylbenzene
SMILES CC1=CC=C(C=C1)I

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 36°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula IC6H4CH3
Quantity 100 g
Beilstein 05, 312
Formula Weight 218.04
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Iodotoluene, 98%
Show More Show Less
Product Identifier
  • 4-Iodotoluene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'