missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropyl-3-methylaniline hydrochloride, 97%, Thermo Scientific™

Product Code. 10304732
Click to view available options
Quantity:
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4534-11-6
Molecular Formula C10H16ClN
Molecular Weight (g/mol) 185.70
MDL Number MFCD00156562
InChI Key VPOAQDSAPBSOLW-UHFFFAOYSA-N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 3-methyl-4-isopropylaniline hydrochloride, 4-isopropyl-3-methylaniline hydrochloride, 4-isopropyl-m-toluidinium chloride, benzenamine, 3-methyl-4-1-methylethyl-, hydrochloride, acmc-20a42e, 3-methyl-4-propan-2-ylaniline hydrochloride, 4-isopropyl-3-methylbenzenamine hydrochloride, 3-methyl-4-methylethyl phenylamine, chloride, 3-methyl-4-propan-2-yl aniline hydrochloride, benzenamine, 3-?methyl-?4-? 1-?methylethyl ?-?, hy
SMILES CC1=C(C=CC(=C1)N)C(C)C.Cl
IUPAC Name 3-methyl-4-propan-2-ylaniline;hydrochloride
PubChem CID 2796270
Formula Weight 185.7
Percent Purity 95%
Chemical Name or Material 4-Isopropyl-3-methylaniline hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.