missing translation for 'onlineSavingsMsg'
Learn More

4'-Isopropylacetophenone, 97%, Thermo Scientific Chemicals

Product Code. 11448977
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-13-6
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.23
MDL Number MFCD00048297
InChI Key PDLCCNYKIIUWHA-UHFFFAOYSA-N
Synonym 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one
PubChem CID 12578
IUPAC Name 1-(4-propan-2-ylphenyl)ethanone
SMILES CC(C)C1=CC=C(C=C1)C(C)=O

Specifications

Density 0.97
Boiling Point 238°C
Refractive Index 1.523
Quantity 5 g
Beilstein 2205694
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 162.23
Percent Purity 98%
Chemical Name or Material 4'-Isopropylacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'