missing translation for 'onlineSavingsMsg'
Learn More

4-(Isopropylcarbamoyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15439697
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397843-67-3
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.036
MDL Number MFCD04039353
InChI Key GBCSEYKTZAKRMT-UHFFFAOYSA-N
Synonym 4-isopropylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenylboronic acid, 4-isopropylcarbamoyl benzeneboronic acid, 4-isopropylcarbamoyl phenylboronic acid, 4-propan-2-ylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenyl boronic acid, n-isopropyl 4-boronobenzamide, boronic acid, 4-1-methylethyl amino carbonyl phenyl, 4-1-methylethyl amino carbonyl phenyl-boronic acid
PubChem CID 2773475
IUPAC Name [4-(propan-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC(C)C)(O)O

Specifications

Melting Point 162°C to 167°C
Quantity 1 g
Formula Weight 207.04
Percent Purity 95%
Chemical Name or Material 4-(Isopropylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'