missing translation for 'onlineSavingsMsg'
Learn More

(4-Isopropylphenoxy)acetyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11347569
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a primary and secondary intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a primary and secondary intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 223128-33-4
Molecular Formula C11H13ClO2
Molecular Weight (g/mol) 212.673
MDL Number MFCD02628421
InChI Key YGVLHWFQKYVLFY-UHFFFAOYSA-N
Synonym 4-isopropylphenoxy acetyl chloride, 2-4-isopropylphenoxy acetyl chloride, acmc-1cokt, 4-iso propyl phenoxy acetyl chloride, 4-isopropyl-phenoxy-acetyl chloride, 4-propan-2-yl phenoxy acetyl chloride, 2-4-methylethyl phenoxy acetyl chloride, 2-4-propan-2-yl phenoxy acetyl chloride, acetyl chloride, 4-1-methylethyl phenoxy, acetyl chloride,2-4-1-methylethyl phenoxy
PubChem CID 21209244
IUPAC Name 2-(4-propan-2-ylphenoxy)acetyl chloride
SMILES CC(C)C1=CC=C(C=C1)OCC(=O)Cl

Specifications

Refractive Index 1.5195
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 212.67
Percent Purity 98%
Chemical Name or Material (4-Isopropylphenoxy)acetyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.