missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylphenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11394454
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1643-16-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00014364
InChI Key FPVCSFOUVDLTDG-UHFFFAOYSA-N
Synonym 4-isopropylphenoxyacetic acid, 2-4-isopropylphenoxy acetic acid, 4-isopropylphenoxy acetic acid, p-i-propylphenoxyacetic acid, 4-isopropyl-phenoxy-acetic acid, 4-propan-2-yl phenoxy acetic acid, 2-4-methylethyl phenoxy acetic acid, 4-isopropylphenoxyaceticacid, akos bbb/199, 2-4-propan-2-ylphenoxy acetic acid
PubChem CID 137131
IUPAC Name 2-(4-propan-2-ylphenoxy)acetic acid
SMILES CC(C)C1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 85°C to 87°C
Quantity 25 g
Beilstein 2261017
Formula Weight 194.23
Percent Purity ≥98%
Chemical Name or Material 4-Isopropylphenoxyacetic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'