missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptobenzoic acid, 90%, Thermo Scientific Chemicals

Product Code. 10264974
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-36-8
Molecular Formula C7H6O2S
Molecular Weight (g/mol) 154.18
MDL Number MFCD00016617
InChI Key LMJXSOYPAOSIPZ-UHFFFAOYSA-N
Synonym 4-mercaptobenzoic acid, 4-mercaptobenzoate, benzoic acid, 4-mercapto, 4-mercapto-benzoic acid, 4-mercapto benzoic acid, 4-thiobenzoic acid, p-mercaptobenzoic acid, benzoic acid, p-mercapto, para-mercaptobenzoate, paracarboxythiophenol
PubChem CID 95738
IUPAC Name 4-sulfanylbenzoic acid
SMILES OC(=O)C1=CC=C(S)C=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 223.0°C to 225.0°C
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Linear Formula HSC6H4CO2H
Quantity 5 g
Formula Weight 154.19
Percent Purity 90%
Chemical Name or Material 4-Mercaptobenzoic acid
compliance-icons
Product Identifier
  • 4-Mercaptobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'