Learn More
4-Methoxy-2-methylphenylboronic acid, 97%, Thermo Scientific Chemicals
59.90 € - 205.00 €
Chemical Identifiers
CAS | 208399-66-0 |
---|---|
Molecular Formula | C8H11BO3 |
Molecular Weight (g/mol) | 165.98 |
MDL Number | MFCD02684315 |
InChI Key | AMSQNQJCBXQYEX-UHFFFAOYSA-N |
Synonym | 4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6 |
PubChem CID | 2773486 |
IUPAC Name | (4-methoxy-2-methylphenyl)boronic acid |
SMILES | COC1=CC=C(B(O)O)C(C)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10297597
|
Thermo Scientific Acros
393850010 |
1 g | Glass Bottle |
59.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10257647
|
Thermo Scientific Acros
393850050 |
5 g | Glass Bottle |
205.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
208399-66-0 | |
165.98 | |
AMSQNQJCBXQYEX-UHFFFAOYSA-N | |
2773486 | |
COC1=CC=C(B(O)O)C(C)=C1 |
C8H11BO3 | |
MFCD02684315 | |
4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6 | |
(4-methoxy-2-methylphenyl)boronic acid |
Specifications
208399-66-0 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD02684315 | |
AMSQNQJCBXQYEX-UHFFFAOYSA-N | |
(4-methoxy-2-methylphenyl)boronic acid | |
2773486 | |
97% | |
4-Methoxy-2-methylphenylboronic acid |
96.0 | |
148.0°C to 161.0°C | |
Authentic | |
C8H11BO3 | |
4-methoxy-2-methylphenyl boronic acid, 2-methyl-4-methoxyphenylboronic acid, 4-methoxy-2-methylbenzeneboronic acid, 4-methoxy-2-methylphenyl boranediol, 4-methoxy-2-methyl-phenyl boronic acid, 4-methoxy-2-methylphenbylboronic acid, 4-methoxy-2-methylphenylboronicacid, boronic acid, 4-methoxy-2-methylphenyl, 2-methyl-4-methoxybenzeneboronic acid, acmc-1cge6 | |
COC1=CC=C(B(O)O)C(C)=C1 | |
165.98 | |
165.98 | |
Crystalline Solid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
- 4-Methoxy-2-methylphenylboronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only