missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide, 97%, Thermo Scientific™

Product Code. 15402576
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346696-69-3
Molecular Formula C15H17NO3S
Molecular Weight (g/mol) 291.365
MDL Number MFCD01213791
InChI Key PAIJRRXAEGQQNV-UHFFFAOYSA-N
Synonym 4-methoxy-n-4-methylbenzyl benzenesulfonamide, 4-methoxy-n-4-methyl-benzyl-benzenesulfonamide, 4-methoxy-n-4-methylphenyl methyl benzenesulfonamide, n-4-methylbenzyl-4-methoxybenzenesulfonamide, 4-methoxyphenyl sulfonyl 4-methylphenyl methyl amine
PubChem CID 743491
IUPAC Name 4-methoxy-N-[(4-methylphenyl)methyl]benzenesulfonamide
SMILES CC1=CC=C(C=C1)CNS(=O)(=O)C2=CC=C(C=C2)OC

Specifications

Quantity 250 mg
Formula Weight 291.36
Percent Purity 97%
Chemical Name or Material 4-Methoxy-N-(4-methylbenzyl)benzenesulfonamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'