missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxyacetophenone, 99%, Thermo Scientific Chemicals

Product Code. 11429093
Click to view available options
Quantity:
25 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4'-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4?-methoxyacetophenone with α,ω-dienes to yield copolymers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methoxyacetophenone is used as a component of perfumes and as chemical intermediate in the manufacture of pharmaceuticals, resins, flavouring agents. And also used to study ruthenium catalyzed step growth copolymerization of 4′-methoxyacetophenone with α,ω-dienes to yield copolymers.

Solubility
Insoluble in water.

Notes
Light Sensitive, Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-06-1
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008745
InChI Key NTPLXRHDUXRPNE-UHFFFAOYSA-N
Synonym 4'-methoxyacetophenone, 4-acetylanisole, 4-methoxyacetophenone, 1-4-methoxyphenyl ethanone, p-methoxyacetophenone, acetanisole, novatone, linarodin, vananote, ethanone, 1-4-methoxyphenyl
PubChem CID 7476
ChEBI CHEBI:86567
IUPAC Name 1-(4-methoxyphenyl)ethanone
SMILES COC1=CC=C(C=C1)C(C)=O

Specifications

Melting Point 36°C to 40°C
Density 1.08
Boiling Point 258°C to 260°C
Flash Point 138°C (280°F)
Odor Characteristic
Quantity 1000 g
Beilstein 742313
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 150.18
Percent Purity 99%
Chemical Name or Material 4'-Methoxyacetophenone
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'