Learn More
4-Methoxybenzaldehyde, 98%, Thermo Scientific Chemicals
Description
4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methoxybenzaldehyde is widely utilized in the fragrance and flavor industry. It finds application as an important intermediate in the synthesis of other organic compounds, perfumes and pharmaceuticals like antihistamines. It is also used in the preparation of agrochemicals, dyes and plastic additives. A solution of para-anisaldehyde with acid and ethanol is used as stain in thin layer chromatography (TLC), which provides easy identification of different compounds.
Solubility
Miscible with acetone, alcohol, ether, chloroform and benzene. Immiscible with water.
Notes
Air and Light Sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
Specifications
Specifications
Melting Point | 0°C to 2°C |
Density | 1.121 |
Boiling Point | 247°C to 249°C |
Flash Point | 116°C (241°F) |
Assay Percent Range | 98% |
Odor | Odorless |
Refractive Index | 1.573 |
Quantity | 50 g |
Beilstein | 471382 |
Sensitivity | Air and light sensitive |
Show More |
Safety and Handling
- 4-Methoxybenzaldehyde
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : p-Anisaldehyde
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.