missing translation for 'onlineSavingsMsg'
Learn More
4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals Product Code.: 15452188

4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals

Product Code. 15452188
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15452188

Brand: Thermo Scientific Alfa Aesar H55827.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.

Solubility
Decompose in water

Notes
Store at -20°C. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 459-64-3
Molecular Formula C7H7BF4N2O
Molecular Weight (g/mol) 221.95
MDL Number MFCD00011897
InChI Key CACFTKIREZJSIG-UHFFFAOYSA-J
Synonym 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide
PubChem CID 517233
SMILES [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N

Specifications

Melting Point 142°C to 144°C
Quantity 25 g
UN Number UN3261
Solubility Information Decompose in water
IUPAC Name boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride
Formula Weight 221.95
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzenediazonium tetrafluoroborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.