missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals

Product Code. 11403017
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.

Solubility
Soluble in water, chloroform, ethyl acetate.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, acid chlorides, acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-13-5
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00004653
InChI Key MSHFRERJPWKJFX-UHFFFAOYSA-N
Synonym 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol
PubChem CID 7738
ChEBI CHEBI:86918
IUPAC Name (4-methoxyphenyl)methanol
SMILES COC1=CC=C(CO)C=C1

Specifications

Melting Point 22°C to 25°C
Density 1.11
Boiling Point 259°C
Flash Point 113°C (235°F)
Refractive Index 1.545
Quantity 100 g
Beilstein 636654
Merck Index 14,604
Solubility Information Soluble in water,chloroform,ethyl acetate.
Formula Weight 138.17
Percent Purity 98%
Chemical Name or Material 4-Methoxybenzyl alcohol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'