missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylchloride, 98%, stabilized, Thermo Scientific Chemicals

Product Code. 10535481
Click to view available options
Quantity:
5 g
25 g
100 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 824-94-2, 584-08-7
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000915
InChI Key MOHYOXXOKFQHDC-UHFFFAOYSA-N
Synonym 4-methoxybenzyl chloride, 4-methoxybenzylchloride, 1-chloromethyl-4-methoxybenzene, p-methoxybenzyl chloride, p-anisyl chloride, alpha-chloro-4-methoxytoluene, p-chloromethyl anisole, benzene, 1-chloromethyl-4-methoxy, 4-chloromethyl anisole, anisole, p-chloromethyl
PubChem CID 69993
IUPAC Name 1-(chloromethyl)-4-methoxybenzene
SMILES COC1=CC=C(C=C1)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -1.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 117.0°C to 118.0°C (14.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490
Quantity 5 g
Specific Gravity 1.15
Solubility Information Solubility in water: slowly decomposes
Stabilizer 1% K2CO3
Formula Weight 156.61
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxybenzylchloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxybenzylchloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'