missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-(Methoxycarbamoyl)benzeneboronic acid, 97%
57.00 €
Chemical Identifiers
CAS | 850568-17-1 |
---|---|
Molecular Formula | C8H10BNO4 |
Molecular Weight (g/mol) | 194.981 |
MDL Number | MFCD04115670 |
InChI Key | MZZWCLBDQROHHS-UHFFFAOYSA-N |
Synonym | 4-o-methylhydroxylaminocarbonyl phenylboronic acid, 4-methoxycarbamoyl phenyl boronic acid, 4-o-methylhydroxylaminocarbonyl benzeneboronic acid, 4-methoxycarbamoyl phenylboronic acid, 4-methoxycarbamoyl benzeneboronic acid, 4-o-methyl hydroxylaminocarbonyl phenylboronic acid, 4-o-methylhydroaminocarbonyl phenylboronic acid |
PubChem CID | 3379984 |
IUPAC Name | [4-(methoxycarbamoyl)phenyl]boronic acid |
SMILES | B(C1=CC=C(C=C1)C(=O)NOC)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15499947
|
Alfa Aesar™
H53188.MD |
250 mg | N/A | ||||||
Description
Chemical Identifiers
850568-17-1 | |
194.981 | |
MZZWCLBDQROHHS-UHFFFAOYSA-N | |
3379984 | |
B(C1=CC=C(C=C1)C(=O)NOC)(O)O |
C8H10BNO4 | |
MFCD04115670 | |
4-o-methylhydroxylaminocarbonyl phenylboronic acid, 4-methoxycarbamoyl phenyl boronic acid, 4-o-methylhydroxylaminocarbonyl benzeneboronic acid, 4-methoxycarbamoyl phenylboronic acid, 4-methoxycarbamoyl benzeneboronic acid, 4-o-methyl hydroxylaminocarbonyl phenylboronic acid, 4-o-methylhydroaminocarbonyl phenylboronic acid | |
[4-(methoxycarbamoyl)phenyl]boronic acid |
Specifications
850568-17-1 | |
C8H10BNO4 | |
4-o-methylhydroxylaminocarbonyl phenylboronic acid, 4-methoxycarbamoyl phenyl boronic acid, 4-o-methylhydroxylaminocarbonyl benzeneboronic acid, 4-methoxycarbamoyl phenylboronic acid, 4-methoxycarbamoyl benzeneboronic acid, 4-o-methyl hydroxylaminocarbonyl phenylboronic acid, 4-o-methylhydroaminocarbonyl phenylboronic acid | |
B(C1=CC=C(C=C1)C(=O)NOC)(O)O | |
194.981 | |
194.98 | |
4-(Methoxycarbamoyl)benzeneboronic acid |
130°C to 136°C | |
MFCD04115670 | |
MZZWCLBDQROHHS-UHFFFAOYSA-N | |
[4-(methoxycarbamoyl)phenyl]boronic acid | |
3379984 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title