missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenyl isothiocyanate, 98%, Thermo Scientific Chemicals

Product Code. 11448236
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenyl isothiocyanate has been used in the synthesis of 3-benzyl-2-(4-methoxyphenyl)-3H-[1,2,4]triazolo-[5,1-b]quinazolin-9-one and 2-{[4-amino-3-(4-methylphenyl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-1-yl]acetyl}-N-arylhydrazinecarbothioamides.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2284-20-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00011676
InChI Key VRPQCVLBOZOYCG-UHFFFAOYSA-N
Synonym 4-methoxyphenyl isothiocyanate, 4-methoxyphenylisothiocyanate, 1-isothiocyanato-4-methoxy-benzene, p-methoxyphenyl isothiocyanate, benzene, 1-isothiocyanato-4-methoxy, isothiocyanic acid 4-methoxyphenyl ester, 4-methoxybenzenisothiocyanate, 4-isothiocyanatoanisole, acmc-1ccqx, p-methoxyphenylisothiocyanate
PubChem CID 75293
IUPAC Name 1-isothiocyanato-4-methoxybenzene
SMILES COC1=CC=C(C=C1)N=C=S

Specifications

Melting Point 18°C to 20°C
Density 1.196
Boiling Point 279°C to 281°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6489
Quantity 10 g
UN Number UN2922
Beilstein 606967
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 165.22
Percent Purity 98%
Chemical Name or Material 4-Methoxyphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'