missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylmagnesium bromide, 0.5M in THF, Thermo Scientific Chemicals

Product Code. 15492997
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13139-86-1
Molecular Formula C7H7BrMgO
Molecular Weight (g/mol) 211.34
MDL Number MFCD00013515
InChI Key JKXAZEXIOPFJSC-UHFFFAOYSA-M
Synonym 4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent
PubChem CID 2734898
SMILES COC1=CC=C([Mg]Br)C=C1

Specifications

Color Brown
Density 0.955 g/mL
Flash Point -20°C (-4°F)
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
IUPAC Name bromo(4-methoxyphenyl)magnesium
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 0.5M in THF
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylmagnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methoxyphenylmagnesium bromide, 0.5M in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, bromo(4-methoxyphenyl)-magnesium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.