Learn More
4-Methoxyphenylmagnesium bromide, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals
82.41 € - 241.20 €
Chemical Identifiers
CAS | 13139-86-1, 109-99-9 |
---|---|
Molecular Formula | C7H7BrMgO |
Molecular Weight (g/mol) | 211.34 |
MDL Number | MFCD00013515 |
InChI Key | JKXAZEXIOPFJSC-UHFFFAOYSA-M |
Synonym | 4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent |
PubChem CID | 2734898 |
IUPAC Name | magnesium;methoxybenzene;bromide |
SMILES | COC1=CC=C([Mg]Br)C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10022442
|
Thermo Scientific Acros
377421000 |
100 mL | AcroSeal™ Glass bottle |
82.41 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10768543
|
Thermo Scientific Acros
377428000 |
800 mL | AcroSeal™ Glass Bottle |
241.20 €
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13139-86-1, 109-99-9 | |
211.34 | |
JKXAZEXIOPFJSC-UHFFFAOYSA-M | |
2734898 | |
COC1=CC=C([Mg]Br)C=C1 |
C7H7BrMgO | |
MFCD00013515 | |
4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent | |
magnesium;methoxybenzene;bromide |
Specifications
13139-86-1, 109-99-9 | |
25.36, 80.98 | |
1.0000g/mL | |
C7H7BrMgO | |
MFCD00013515 | |
4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent | |
JKXAZEXIOPFJSC-UHFFFAOYSA-M | |
magnesium;methoxybenzene;bromide | |
2734898 | |
Liquid |
19.02, 74.64 | |
Yellow to Brown | |
−21°C | |
CH3O(C6H4)MgBr | |
1 | |
Solubility in water: vigorous reaction | |
COC1=CC=C([Mg]Br)C=C1 | |
211.34 | |
211.34 | |
4-Methoxyphenylmagnesium bromide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
Recommended Storage : Magnesium salts may crystallize below 20°C
- 4-Methoxyphenylmagnesium bromide, 1M solution in THF
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only