missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methyl-1,2,3-thiadiazole-5-carbonyl chloride, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
IUPAC Name | 4-methyl-1,2,3-thiadiazole-5-carbonyl chloride |
Formula Weight | 162.6 |
Percent Purity | 90% |
Chemical Name or Material | 4-Methyl-1, 2, 3-thiadiazole-5-carbonyl chloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction